Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-20 R beta Fc Chimera Protein

Catalog No. 1788IR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1788IR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1788IR050 Supplier R&D Systems™ Supplier No. 1788IR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-20 R beta Fc Chimera Protein is derived from NS0. The Recombinant Human IL-20 R beta Fc Chimera Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 53833
Molecular Weight (g/mol) 49.2 kDa (monomer)
Name IL-20 R beta Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human IL-20 R beta protein, (N-terminus) Human IL-20 R beta (Asp30-Ala230) Accession &Num; Q6UXL0 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.