Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-22BP His-tag Protein

Catalog No. 8498BP025CF
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8498BP025CF 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8498BP025CF Supplier R&D Systems™ Supplier No. 8498BP025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-22BP His-tag Protein is derived from CHO. The Recombinant Human IL-22BP His-tag Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_851826
For Use With (Application) Bioactivity
Gene ID (Entrez) 116379
Molecular Weight (g/mol) Observed M.W.: 46-53kDa, reducing conditions, Theoretical M.W.: 26kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human IL-22BP protein Thr22-Pro231,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16
Common Name IL-22BP/IL22 RA2
Gene Symbol IL22RA2
Biological Activity Measured by its ability to inhibit IL-22-induced IL-10 secretion by COLO 205 human colorectal adenocarcinoma cells. The ED50 for this effect is typically 1-5ng/mL in the presence of 1ng/mL of Recombinant Human IL-22.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Thr22-Pro231, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.