Learn More
R&D Systems™ Recombinant Human IL-23 (HEK293 Expressed) Protein
Description
Specifications
Specifications
| Accession Number | P29460.1 & AAG37232.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 51561 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 57-71 kDa, under reducing conditions., MolecularWeight-theroretical: 55 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human IL-23 protein Human IL-12p40 (Ile23-Ser328) (N-terminus) Accession # P29460.1 GSGSSRGGSGSGGSGGGGSKL Human IL-23p19 (Arg20-Pro189) Accession # AAG37232.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | IL23, IL-23, IL-23 p19/IL-12 p40, IL-23 A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.