Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-23 Protein
GSA_VASDP

Catalog No. 1290IL010 Shop All Novus Biologicals Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in MES, NaCl and EDTA with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in MES, NaCl and EDTA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
1290IL010 Lyophilized from a 0.2μm filtered solution in MES, NaCl and EDTA with BSA as a carrier protein.
1290IL010CF Supplied as a 0.2μm filtered solution in MES, NaCl and EDTA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 1290IL010 Supplier R&D Systems™ Supplier No. 1290IL010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-23 Protein is derived from Sf 21 (baculovirus). The Recombinant Human IL-23 Protein has been validated for the following applications: Bioactivity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in MES, NaCl and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 51561
Molecular Weight (g/mol) 55.0 kDa (IL-23 single chain); 36.5 kDa (p40) & 18.5 kDa (p19)
Name IL-23
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-23 protein, (N-terminus) Human IL-23 p40 (Ile23-Ser328) Accession &Num; P29460 GSGSSRGGSGSGGSGGGGSKL Human IL-23 p19 (Arg20-Pro189) Accession &Num; AAG37232 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.