Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-23 Protein
SDP

Catalog No. 1290IL500CF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-90I-L500CF 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-90I-L500CF Supplier R&D Systems™ Supplier No. 1290IL500/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-23 Protein is derived from Sf 21 (baculovirus). The Recombinant Human IL-23 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P29460 (p40) and AAG37232(p19)
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2μm filtered solution in MES, NaCl and EDTA.
Gene ID (Entrez) 51561
Molecular Weight (g/mol) 55.0kDa (IL-23 single chain), 36.5kDa (p40), 18.5kDa (p19)
Quantity 500 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-23 protein, (N-terminus) Human IL-23 p40 (Ile23-Ser328) Accession &Num; P29460 GSGSSRGGSGSGGSGGGGSKL Human IL-23 p19 (Arg20-Pro189) Accession &Num; AAG37232 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0EU per 1μg of the protein by the LAL method.
Gene Symbol IL23A
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.