Learn More
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 282618 |
| Molecular Weight (g/mol) | 21.4 kDa |
| Name | IL-29/IFN-lambda 1 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human IL-29/IFN-lambda 1 protein Gly20-Thr200,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

