Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-3 Protein

Catalog No. 203IL100
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
203IL100 100 μg Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein.
203IL010 10 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
203IL050 50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
203IL01M 1 mg Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein.
203IL250 250 μg Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein.
203IL500 500 μg Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 203IL100 Supplier R&D Systems™ Supplier No. 203IL100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.02-0.1 ng/mL. The specific activity of Recombinant Human IL-3 is approximately 1.8 x 103 IU/μg, which is calibrated against recombinant human IL-3 WHO International Standard (NIBSC code: 91/510). Specific activity is for reference purposes only and is not routinely tested.

Specifications

Accession Number AAC08706
Formulation Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 3562
Molecular Weight (g/mol) 15 kDa
Name IL-3
Quantity 100 μg
Source E. coli-derived human IL-3 protein Ala20-Phe152,with and without an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.