Learn More
R&D Systems™ Recombinant Human IL-3 Protein
Description
Specifications
Specifications
| Accession Number | AAC08706 |
| Formulation | Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 3562 |
| Molecular Weight (g/mol) | 15 kDa |
| Name | IL-3 |
| Quantity | 500 μg |
| Source | E. coli-derived human IL-3 protein Ala20-Phe152,with and without an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.