Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IL-31 Animal-Free Protein
SDP

Catalog No. NBP2761955U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
25 μg
5 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2761955U 5 μg
NB27619525U 25 μg
NBP2761951M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP2761955U Supplier Novus Biologicals™ Supplier No. NBP2761955UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • An un-tagged bioactive recombinant protein corresponding to Human IL-31
  • Amino Acid Sequence: SHTLPVRLLR PSDDVQKIVE ELQSLSKMLL KDVEEEKGVL VSQNYTLPCL SPDAQPPNNI HSPAIRAYLK TIRQLDNKSV IDEIIEHLDK LIFQDAPETN ISVPTDTHEC KRFILTISQQ FSECMDLALK SLTSGAQQAT T
  • Produced with no materials of animal or human origin using ANIMAL-FREE purification processes.
  • No animal-derived ingredients, No FBS
  • E. coli based production system
  • Animal-Free facility, fermentor, columns, filters, labware, lyophilization, and storage
  • Endotoxin levels at <1 EUs/ug
  • Purity levels at 95% or greater

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Gene ID (Entrez) 386653
Molecular Weight (g/mol) 15.8 kDa
Purification Method >95%, by SDS-PAGE
Quantity 5 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Gene Symbol IL31
Specific Reactivity IL-31 is a monomer, 15.8 kDa (141 aa)
Reconstitution Sterile water at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 20 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.