Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-31 (HEK293-expressed) Protein
SDP

Catalog No. 10425IL020 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10425IL020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10425IL020 Supplier R&D Systems™ Supplier No. 10425IL020

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Recombinant Human IL-31 (HEK293-expressed) (Catalog # 10425-IL) stimulates CCL2/JE/MCP-1 secretion from the BEAS-2B Human lung bronchial epithelial cell line. The ED50 for this effect is <5 ng/mL.

Specifications

Accession Number Q6EBC2.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 386653
Molecular Weight (g/mol) M.W.-observed: 19-29 kDa, under reducing conditions, M.W.-theoretical: 16 kDa
Quantity 20 μg
Source Human embryonic kidney cell, HEK293-derived human IL-31 protein Leu27-Thr164
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IL31, IL-31 interleukin-31, interleukin 31
Conjugate Unconjugated
Reconstitution Reconstitute at 200 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-31
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.