Learn More
Enzo Life Sciences IL-35 (human) : Fc (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences ALX522140C010
Encompass_Preferred
Highly pure interleukin involved in T cell regulation. Alternative name: Interleukin-35, EBI3 : IL-12A, IL-27B : CLMF p35. MW: ~85kDa. Concentration: 0.1mg/ml after reconstitution. Endotoxin content: <0.1EU/µg purified protein (LAL test; Associates of Cape Cod). Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. Human EBI3 (aa 21-229) linked to human IL-12A (aa 23-219) via the linker GGGSGGGSGGGSGGGS is fused at the C-terminus to the Fc portion of human IgG1 and a FLAG-tag. Use/Stability: Stable for at least 6 months after receipt when stored at -20°C. UniProt: Q14213 (IL-27B), P29459 (IL-21A). Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C. Long Term Storage: -20°C
Catalog No. NC0064105
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.