Learn More
R&D Systems™ Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with BSA as a carrier protein. |
| Gene ID (Entrez) | 27179 |
| Molecular Weight (g/mol) | 17 kDa |
| Name | IL-36 alpha/IL-1F6 (aa 6-158) |
| Quantity | 10 μg |
| Source | E. coli-derived human IL-36 alpha/IL-1F6 protein Lys6-Phe158 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Monomer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.