Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein

Catalog No. 6995IL010CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with Trehalose.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
6995IL010CF Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with Trehalose.
6995IL010 Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 6995IL010CF Supplier R&D Systems™ Supplier No. 6995IL010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein is derived from E. coli. The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with Trehalose.
Gene ID (Entrez) 27179
Molecular Weight (g/mol) 17 kDa
Name IL-36 alpha/IL-1F6 (aa 6-158)
Quantity 10 μg
Source E. coli-derived human IL-36 alpha/IL-1F6 protein Lys6-Phe158
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Monomer
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100μg/mL in Water.
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.