Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-36 beta/IL-1F8 (aa 5-157) Protein

Catalog No. 6834ILB025C
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-34I-LB025C 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 68-34I-LB025C Supplier R&D Systems™ Supplier No. 6834ILB025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-36 beta/IL-1F8 (aa 5-157) Protein is derived from E. coli. The Recombinant Human IL-36 beta/IL-1F8 (aa 5-157) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_775270
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 27177
Molecular Weight (g/mol) Observed M.W.: 17kDa, reducing conditions, Theoretical M.W.: 17kDa
Quantity 25 μg
Source E. coli-derived human IL-36 beta/IL-1F8 protein Arg5-Glu157
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias family of interleukin 1-eta, FIL1, FIL1 eta, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1 eta, IL1-ETA, IL1F8, IL-1F8, IL1F8 (Canonical product IL-1F8a), IL-1F8 (FIL1-eta), IL1H2, IL-1H2, IL36 beta, IL36B, interleukin 1 family, member 8 (eta), interleukin 1, eta, Interleukin 36, Beta, Interleukin-1 eta, interleukin-1 family member 8, Interleukin-1 homolog 2, Interleukin-1 Superfamily e, Interleukin-36 Beta
Common Name IL-36 beta/IL-1F8
Gene Symbol IL36B
Biological Activity Measured by its ability to induce IL-8 secretion in A431 human epithelial carcinoma cells. The ED50 for this effect is typically 0.8-4.8ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Arg5-Glu157
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.