Learn More
Description
Specifications
Specifications
| Accession Number | Q14213, Q9NPF7 |
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | 69 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell,HEK293-derived human IL-39 (IL-23p19/EBI3) protein Human EBI3 (Arg21-Lys229,Gln95Cys) Accession &Num; Q14213 GGGSGGGSGGGS Human IL-23p19 (Arg20-Pro189) Accession &Num; Q9NPF7 IEGRMD Human IgG1 (Pro100-Lys330) |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Specific Reactivity | IL-39 (IL-23p19/EBI3) |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
