Learn More
Proteintech Group Inc Recombinant Human IL-6 1000ug
SDP
Supplier:  Proteintech Group Inc HZ10191000UG
Encompass_Preferred
Animal-free Recombinant Human IL-6 is produced in human 293 cells as a monomeric glycoprotein with an apparent molecular mass of 21 to 29 kDa. This broad molecular mass is attributable to glycosylation, which is absent when this cytokine is expressed in E. coli. Glycosylation contributes to stability in cell growth media and other applications. IL-6 is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses. Produced by T cells, monocytes, fibroblasts, endothelial cells, and keratinocytes, IL-6 has diverse biological functions.
Catalog No. 50-197-3852
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.