Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-6 Protein
GSA_VA

Catalog No. 206IL200CF
Change view
Click to view available options
Quantity:
1 mg
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-6IL-200CF 200 μg
206IL001MGC 1 mg
206IL050CF 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 20-6IL-200CF Supplier R&D Systems™ Supplier No. 206IL200/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

A New rh IL-6 is Available! The new protein has ∼2 fold better activity and is HEK293 expressed!

Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.2-0.8 ng/mL.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accession Number Q75MH2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
Gene ID (Entrez) 3569
Molecular Weight (g/mol) M.W. theroretical: 20.9 kDa
Name IL-6
Quantity 200 μg
Source E. coli-derived human IL-6 protein Pro29-Met212
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Gene Symbol IL6
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100-200 μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein IL-6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.