Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-7 GMP Protein, CF
SDP

Catalog No. BT007GMP025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT007GMP025 25 μg
BT007GMP010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BT007GMP025 Supplier R&D Systems™ Supplier No. BT007GMP025
Add to Cart
Add to Cart

Animal-Free.

Measured in a cell proliferation assay using PHA-activated human peripheral blood lymphocytes (PBL). Yokota, T. et al. (1986) Proc. Natl. Acad. Sci. USA 83:5894. The ED50 for this effect is 0.100-0.500 ng/mL.The specific activity of Recombinant Human IL-7 is >1.00 x 108 units/mg, which is calibrated against the human IL-7 reference standard (NIBSC code: 90/530).

Specifications

Accession Number P13232.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3574
Molecular Weight (g/mol) Observed: 17 kDa, under reducing conditions, Observed: 17 kDa
Quantity 25 μg
Source E. coli-derived human IL-7 protein Asp26-His177, with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidelines.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 12 months when stored at ≤ -20°C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Regulatory Status GMP
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias IL7, IL-7interleukin-7, interleukin 7, Lymphopoietin-1, PBGF
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100-500 μg/mL in PBS.
Purity or Quality Grade >97%, by SDS-PAGE with quantitative densitometry by Coomassie™ Blue Staining. The molecular weight by mass spectrometry is 17507 Da ± 50 Da
Protein IL-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.