Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-7 (HEK-293-expressed) Protein
SDP

Catalog No. 11089IL010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11089IL010 10 μg
11089IL01M 1 mg
11089IL050 50 μg
11089IL250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11089IL010 Supplier R&D Systems™ Supplier No. 11089IL010
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using PHA-activated human peripheral blood lymphocytes (PBL). Yokota, T. et al. (1986) Proc. Natl. Acad. Sci. USA 83:5894. The ED50 for this effect is 0.100-0.500 ng/mL. The specific activity of Recombinant Human IL-7 is >1.00 x 108 units/mg, which is calibrated against the human IL-7 reference standard (NIBSC code: 90/530). Specific activity is for reference purposes only and is not routinely tested.

Specifications

Accession Number P13232.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3574
Molecular Weight (g/mol) MolecularWeight-observed: 18-28 kDa, under reducing conditions.; MolecularWeight-theroretical: 17 kDa
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived human IL-7 protein Asp26 - His177
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IL7, IL-7 interleukin-7, interleukin 7, Lymphopoietin-1, PBGF
Conjugate Unconjugated
Reconstitution Reconstitute at 100 - 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.