Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-7 Protein
SDP

Catalog No. 207IL005CF Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
200 μg
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
207IL005CF 5 μg Lyophilized from a 0.2μm filtered solution in PBS.
207IL025 25 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
207IL005 5 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
207IL025CF 25 μg Lyophilized from a 0.2μm filtered solution in PBS.
20-7IL-200 200 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
20-7IL-200CF 200 μg Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 207IL005CF Supplier R&D Systems™ Supplier No. 207IL005/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using PHA-activated human peripheral blood lymphocytes (PBL). Yokota, T. et al. (1986) Proc. Natl. Acad. Sci. USA 83:5894. The ED50 for this effect is 0.1-0.5 ng/mL. The specific activity of Recombinant Human IL-7 is approximately 4.4 x 105 IU/μg, which is calibrated against human IL-7 WHO International Standard (NIBSC code: 90/530). Specific activity is for reference purposes only and is not routinely tested.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3574
Molecular Weight (g/mol) 17 kDa
Name IL-7
Quantity 5 μg
Source E. coli-derived human IL-7 protein Asp26-His177,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute 5 μg vials at 50 μg/mL in sterile PBS. Reconstitute 10 μg or larger vials (at 100 μg/mL in sterile PBS.)
Buffer Lyophilized from a 0.2μm filtered solution in PBS.
Product Line R&D Systems
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.