Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-7 Protein

Catalog No. 207IL010
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-7IL0-10 10 μg
20-7IL0-50 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-7IL0-10 Supplier R&D Systems™ Supplier No. 207IL010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using PHA-activated human peripheral blood lymphocytes (PBL). Yokota, T. et al. (1986) Proc. Natl. Acad. Sci. USA 83:5894. The ED50 for this effect is 0.1-0.5 ng/mL.

Specifications

Accession Number P13232
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 3574
Molecular Weight (g/mol) 17 kDa
Quantity 10 μg
Source <EM>E. coli</EM>-derived human IL-7 protein Asp26-His177, with an N-terminal Met
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <0.01 EU per 1 μg of the protein by the LAL method.
Gene Symbol IL7
Conjugate Unconjugated
Cross Reactivity IL-7
Species Human
Reconstitution Reconstitute at 50 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.