Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-8/CXCL8 Protein

Catalog No. 208IL050
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in deionized water with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in deionized water.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
208IL050 50 μg Lyophilized from a 0.2μm filtered solution in deionized water with BSA as a carrier protein.
208IL010 10 μg Lyophilized from a 0.2μm filtered solution in deionized water with BSA as a carrier protein.
208IL010CF 10 μg Lyophilized from a 0.2μm filtered solution in deionized water.
208IL050CF 50 μg Lyophilized from a 0.2μm filtered solution in deionized water.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 208IL050 Supplier R&D Systems™ Supplier No. 208IL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-8/CXCL8 Protein is derived from E. coli. The Recombinant Human IL-8/CXCL8 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in deionized water with BSA as a carrier protein.
Gene ID (Entrez) 3576
Molecular Weight (g/mol) 8 kDa
Name CXCL8/IL-8
Quantity 50 μg
Source E. coli-derived human IL-8/CXCL8 protein Ser28-Ser99
Storage Requirements 12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution.
Gene Alias 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.