Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-9 Protein

Catalog No. 209ILB010CF
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-9IL-B010CF 10 μg
20-9IL-B050CF 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-9IL-B010CF Supplier R&D Systems™ Supplier No. 209ILB010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-9 Protein is derived from Sf 21 (stably transfected). The Recombinant Human IL-9 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P15248
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 3578
Molecular Weight (g/mol) 12 to 24 kDa
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (stably transfected)-derived human IL-9 protein Gln19-Ile144
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name Cytokine P40; HP40; IL9; IL-9; IL-9homolog of mouse T cell and mast cell growth factor 40; interleukin 9; interleukin-9; p40 cytokine; p40 T-cell and mast cell growth factor; P40; T-cell growth factor P40
Biological Activity 0.03 to 0.3ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.