Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL1RAPL2 His-tag Protein

Catalog No. 11207MR100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11207MR100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11207MR100 Supplier R&D Systems™ Supplier No. 11207MR100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA.

When Recombinant Human IL1RAPL2 His-tag (Catalog # 11207-MR) is immobilized at 2 μg/mL (100 μL/well), Recombinant Human PTPRD Fc Chimera (Catalog # 9995-PR ) binds with an ED50 of 45.0-450 ng/mL.

Specifications

Accession Number Q9NP60.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 26280
Molecular Weight (g/mol) M.W.-observed: 40-60 kDa, under reducing conditions., M.W.-theroretical: 40 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IL1RAPL2 protein Thr17-Glu356, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IL-1R10, IL-1 R-9, IL1R9IL-1 receptor accessory protein-like 2, IL-1R9IL-1 RAPL-2, IL1RAPL2, IL-1 RAPL2, IL-1-RAPL-2, IL1RAPL-2 IL1RAPL-2-related protein, interleukin 1 receptor 9, interleukin 1 receptor accessory protein-like 2, Three immunoglobulin domain-containing IL-1 receptor-related 1, TIGIRR-1, TIGIRR-1 Interleukin-1 receptor 9, X-linked interleukin-1 receptor accessory protein-like 2
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL1RAPL2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.