Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ILDR1 Fc Chimera Protein
SDP

Catalog No. 10152D1050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-152-D1050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-152-D1050 Supplier R&D Systems™ Supplier No. 10152D1050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Immunoglobulin-like domain containing receptor 1, or ILDR1, is a single pass Type I membrane protein that belongs to the immunoglobulin superfamily

  • Recombinant Protein
  • Carrier Free (CF)
  • The carrier free version does not contain Bovine Serum Albumin (BSA)

Specifications

Accession Number Q86SU0
For Use With (Application) Bioactivity
Gene ID (Entrez) 286676
Molecular Weight (g/mol) 43 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human ILDR1 protein

Human ILDR1 (Leu24-His167) Accession # Q86SU0IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after opening. 3 months, -20 to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 200 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.