Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IMP Dehydrogenase 2/IMPDH2 Protein

Catalog No. 8349DH050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-49D-H050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-49D-H050 Supplier R&D Systems™ Supplier No. 8349DH050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IMP Dehydrogenase 2/IMPDH2 Protein is derived from Sf 21 (baculovirus). The Recombinant Human IMP Dehydrogenase 2/IMPDH2 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P12268
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, DTT and Glycerol.
Gene ID (Entrez) 3615
Molecular Weight (g/mol) 57 kDa
Quantity 50 μg
Source <EM>Spodoptera frμgiperda</EM>, <EM>Sf</EM> 21 (baculovirus)-derived human IMP Dehydrogenase 2/IMPDH2 protein Met1-Phe514, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles; 6 months from date of receipt, -20 to -70°C as supplied; 3 months, -20 to -70°C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity IMP Dehydrogenase 2/IMPDH2
Species Human
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.