Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Inhibin A Protein

Catalog No. 8506AB010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-06A-B010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-06A-B010 Supplier R&D Systems™ Supplier No. 8506AB010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Inhibin A Protein is derived from CHO. The Recombinant Human Inhibin A Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P05111 (alpha chain) & P08476 (beta chain)
For Use With (Application) Bioactivity
Gene ID (Entrez) 3623
Molecular Weight (g/mol) Observed M.W.: 12-25kDa, reducing conditions, Theoretical M.W.: 15kDa (alpha chain) & 13kDa (beta chain)
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Inhibin A protein Ser233-Ile366 (alpha chain) & Gly311-Ser426 (beta chain)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias A-inhibin subunit, INHA, Inhibin A, inhibin alpha chain, inhibin, alpha
Common Name Inhibin alpha
Gene Symbol INHA
Biological Activity Measured by its ability to neutralize Activin-mediated erythroid differentiation of K562 human chronic myelogenous leukemia cells. The ED50 for this effect is typically 3-18ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ser233-Ile366 (alpha chain) & Gly311-Ser426 (beta chain)
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.