Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human INSRR Protein

Catalog No. 9566RR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9566RR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9566RR050 Supplier R&D Systems™ Supplier No. 9566RR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Human INSRR Protein CF contains an extracellular alpha subunit and an extracellular/transmembrane/cytoplasmic beta subunit.

The Recombinant Human INSRR Protein is derived from CHO.

Specifications

Accession Number P14616
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3645
Molecular Weight (g/mol) 100kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human INSRR protein, (N-terminus) Human INSRR beta (Ala747-Leu923) Accession &Num; P14616 HHHHHH Human INSRR alpha (Leu24-Gly742) Accession &Num; P14616 Human INSRR alpha (Leu24-Gly742) Accession &Num; P14616 Human INSRR beta (Ala747-Leu923) Accession &Num; P14616 HHHHHH (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0EU per 1μg of the protein by the LAL method.
Gene Alias EC 2.7.10, EC 2.7.10.1, insulin receptor-related protein, insulin receptor-related receptor, IRR, IRRIR-related receptor
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.