Learn More
Creative Biomart Insulin Lispro(Recombinant)
Supplier: Creative Biomart THP0130
The product is a recombinant human insulin analogue produced in a specialized laboratory strain of Escherischia coli. Plasmid DNA transfected into the bacteria encodes for an analogue of human insulin that has a lysine at residuce B28 and proline at B29; these residues are reversed in endogenous human insulin. Reversal of these amino acid residues produces a rapid-acting insulin analogue.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.