Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3672 |
| Molecular Weight (g/mol) | 132.3 kDa (Integrin alpha1) & 86.4 kDa (Integrin beta1) |
| Name | Integrin alpha 1 beta 1 |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 1 beta 1 protein, (N-terminus) Human Integrin alpha 1 (Met1-Pro1141) Accession &Num; P56199 GGGSGGGSGGGS Acidic Tail HHHHHH Human Integrin beta 1 (Gln21-Asp728) Accession &Num; P05556 GGGSGGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
