Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Integrin alpha 10 beta 1 Protein

Catalog No. 5895AB050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5895AB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5895AB050 Supplier R&D Systems™ Supplier No. 5895AB050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Mouse Laminin I (Catalog # 3400-010-02 ) is coated at 10 μg/mL, Recombinant Human Integrin alpha 10 beta 1 binds with an apparent Kd <5 nM.

Specifications

Accession Number O75578 (Integrin alpha 10) & P05556 (Integrin beta 1)
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8515
Molecular Weight (g/mol) M.W. (Observed): 160-175 kDa & 125-150 kDa, reducing conditions; M.W. (theoretical): 128.8 kDa (alpha 10) & 86.4 kDa (beta 1)
Name Integrin alpha 10 beta 1
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Integrin alpha 10 beta 1 protein Human Integrin alpha 10 (Phe23-Ser1122) (N-terminus) Accession # O75578GGGSGGGSGGGS Acidic Tail Human Integrin ss1 (Gln21-Asp728) (N-terminus) Accession # P05556 GGGSGGGSGGGS Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Integrin alpha 10 beta 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.