Learn More
R&D Systems™ Recombinant Human Integrin alpha 2b beta 3 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3674 |
| Molecular Weight (g/mol) | 113.6 kDa (Integrin alpha2b) & 84.8 kDa (Integrin beta3) |
| Name | Integrin alpha 2b beta 3 |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 2b beta 3 protein, (N-terminus) Human Integrin alpha 2b Met1-Arg993 (R887L) Accession &Num; P08514 GGGSGGGSGGGS Acidic Tail HHHHHH Human Integrin beta 3 (Gly27-Asp718) Accession &Num; AAA52589 HP GGGSGGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.