Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and MgCl 2 . |
| Gene ID (Entrez) | 3684 |
| Molecular Weight (g/mol) | 124.7 kDa (Integrin alphaM), 79 kDa (Integrin beta2) |
| Name | Integrin alpha M beta 2 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha M beta 2 protein, (N-terminus) Human Integrin alpha M (Phe17-Asn1105) Accession &Num; NP_001139280 acidic tail Human Integrin beta 2 (Gln23-Asn700) Accession &Num; P05107 basic tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
