Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 3685 |
| Molecular Weight (g/mol) | 110.5 kDa ( alphaV subunit), 78.6 kDa (beta6 subunit) |
| Name | Integrin alpha V beta 6 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha V beta 6 protein, (N-terminus) Human Integrin alpha V (Phe31-Val992) Accession &Num; NP_002201.1 His-Pro GGGSGGGS Acidic Tail Human Integrin beta 6 (Gly22-Asn707) Accession &Num; P18564 His-Pro GGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
