Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Integrin alpha V beta 8 Protein

Catalog No. 4135AV050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4135AV050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4135AV050 Supplier R&D Systems™ Supplier No. 4135AV050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human Integrin alpha V beta 8 at 2 μg/mL can bind recombinant human Lap with an apparent Kd <0.5 nM.

Specifications

Accession Number NP_002201.1 (Integrin alpha V) & P26012.1 (Integrin beta 8)
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and MgCl2.
Gene ID (Entrez) 3685
Molecular Weight (g/mol) M.W. (Observed): 130-155 kDa and 85-100 kDa, reducing conditions; M.W. (theoretical): 110.5 kDa (alpha V subunit), 75.3 kDa (beta 8 subunit)
Name Integrin alpha V beta 8
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Integrin alpha V beta 8 protein Human Integrin alpha V (Phe31-Val992) (N-terminus) Accession # NP_002201.1 His-Pro GGGSGGGS Acidic Tail Human Integrin beta 8 (Glu43-Arg684) (N-terminus) Accession # P26012.1 His-Pro GGGSGGGS Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 2 weeks, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol ITGAV
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Integrin alpha V beta 8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.