Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Integrin alpha X beta 2 Protein

Catalog No. 5755AX050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5755AX050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5755AX050 Supplier R&D Systems™ Supplier No. 5755AX050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Integrin alpha X beta 2 Protein is derived from CHO. The Recombinant Human Integrin alpha X beta 2 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3687
Molecular Weight (g/mol) 128.9 kDa ( alphaX) & 83.1 kDa (beta2)
Name Integrin alpha X beta 2
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha X beta 2 protein, (N-terminus) Human Integrin alpha X (Phe20-Pro1107) Accession &Num; P20702 Acidic Tail 6-His tag Human Integrin beta 2 (Gln23-Asn700) Accession &Num; AAA59490 Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.