Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Recombinant Human Interleukin-7

Catalog No. 14561352 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-561-352 0.025 mg
14-561-362 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-561-352 Supplier Sartorius Supplier No. CYK00251008
Only null left
Add to Cart
Add to Cart

RUO Growth Factors and Cytokines

  • Research Use Only
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Organoid Expansion
Quantity 0.025 mg
Product Type Recombinant Proteins
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.