Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IPP Protein
SDP

Catalog No. NBC118418 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118418 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118418 Supplier Novus Biologicals™ Supplier No. NBC118418
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-157 of Human E.coli IPP The Recombinant Human IPP Protein is derived from E. coli. The Recombinant Human IPP Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_005888
For Use With (Application) ELISA, SDS-PAGE
Formulation 10mM HEPES buffer (pH7.4), 25mM NaCl
Gene ID (Entrez) 3652
Molecular Weight (g/mol) 17.3kDa
Name IPP Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MANEDCPKAADSPFSSDKHAQLILAQINKMRNGQHFCDVQ502510155025101514%SDS-PAGELQVGQESFKAHRLVLAASSPYFAALFTGGMKESSKDVVPILGIEAGIFQILLDFIYTGIVNIGVNNVQELIIAADMLQLTEVVHLCCEFLKGQIDPLNCIGIFQFSEQIACHDLLEF
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Research Category Cell Cycle and Replication
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.