Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Kallikrein 5 His Protein
SDP

Catalog No. NBP19890520 Shop All R&D Systems Products
Change view
Click to view available options
20μg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP19890520 20μg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP19890520 Supplier Novus Biologicals™ Supplier No. NBP19890520UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 67-293 of Human Kallikrein 5 The Recombinant Human Kallikrein 5 Protein is derived from E. coli. The Recombinant Human Kallikrein 5 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q9Y337
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 2M Urea, 20% glycerol
Gene ID (Entrez) 25818
Name Kallikrein 5 Protein
Purification Method This product is useful for SDS-PAGE. The purity of this protein is >90% by SDS - PAGE.
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.