Learn More
Description
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.4M UREA, 10% glycerol |
| Gene ID (Entrez) | 27094 |
| Molecular Weight (g/mol) | TMW: 16.8kDa |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | BK channel subunit beta-3, BKbeta3, HBETA3, KCNMB2, KCNMBL, potassium large conductance calcium-activated channel, subfamily M beta member3, slo-beta-3, subfamily M subunit beta-3, voltage and Ca2+ activated potassium channel Maxi K beta 3subunit |
| Common Name | KCNMB3 |
| Conjugate | Unconjugated |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
