Learn More
R&D Systems™ Recombinant Human KEAP1/CUL3/NEDD8/RBX1 Complex Protein, CF
Description
Specifications
Specifications
| Accession Number | Q14145, Q13618, Q15843, P62877 |
| For Use With (Application) | Bioactivity |
| Formulation | X mg/ml (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM DTT |
| Molecular Weight (g/mol) | M.W.-theroretical: 70 kDa (KEAP1), 101 kDa (neddylated CUL3), 12 kDa (RB x 1) |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human KEAP1/CUL3/NEDD8/RBX1 protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 48 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
| Protein | KEAP1/CUL3/NEDD8/RBX1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.