Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human KGF/FGF-7 Protein
SDP

Catalog No. 251KG01M Shop All Novus Biologicals Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
25-1KG-01M Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA with BSA as a carrier protein.
25-1KG-01MCF Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-1KG-01M Supplier R&D Systems™ Supplier No. 251KG01M

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using 4MBr-5 rhesus monkey epithelial cells. Rubin, J.S. et al. (1989) Proc. Natl. Acad. Sci. USA 86:802. The ED50 for this effect is typically 6-60 ng/mL.

Specifications

Accession Number P21781
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 2252
Molecular Weight (g/mol) 19kDa
Quantity 1 mg
Source E. coli-derived human KGF/FGF-7 protein Cys32-Thr194,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor)
Gene Symbol FGF7
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.