Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human KGF/FGF-7 Protein

Catalog No. 11340KG500
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-340-KG500 500 μg
11-340-KG100 100 μg
11-340-KG050 50 μg
11-340-KG01M 1 mg
11-340-KG010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 11-340-KG500 Supplier R&D Systems™ Supplier No. 11340KG500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is 25.0-250 ng/mL.

Specifications

Accession Number P21781.1
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with Trehalose.
Gene ID (Entrez) 2252
Molecular Weight (g/mol) MolecularWeight-observed: 22-28 kDa, under reducing conditions., MolecularWeight-theroretical: 19 kDa
Quantity 500 μg
Source Human embryonic kidney cell, HEK293-derived human KGF/FGF-7 protein Cys32-Thr194
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias FGF7, FGF-7, fibroblast growth factor 7 HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor)
Conjugate Unconjugated
Reconstitution Reconstitute 10 μg size at 100 μg/mL and all the other sizes (at 250 μg/mL in PBS.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein KGF/FGF-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.