Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Sodium Acetate and NaCl. |
| Gene ID (Entrez) | 3827 |
| Molecular Weight (g/mol) | 71 kDa (mature), 41 kDa (heavy) & 30 kDa (light) |
| Name | Kininogen High Molecular Weight (HKa) |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Kininogen protein Gln19-Ser644,with an N-terminal signal sequence and a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | High molecular weight form consisting of the mature chain, the heavy and light chains, and a minor species |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
