Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Klotho His-tag Protein

Catalog No. 10308KL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-308-KL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-308-KL050 Supplier R&D Systems™ Supplier No. 10308KL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mucin Stalk Chimera

Measured by its binding ability in a functional ELISA. When Recombinant Human FGF-23 (Catalog # 2604-FG ) is immobilized at 5 μg/mL (100 μL/well), Recombinant Human Klotho Mucin Stalk Chimera His-tag (Catalog # 10308-KL) binds with an ED50 of 0.2-1.8 μg/mL.

Specifications

Accession Number Q9UEF7 (Klotho) & P78423 (Fractalkine Mucin-like Stalk)
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 9365
Molecular Weight (g/mol) M.W. (Observed): 110-140 kDa, under reducing conditions; M.W. (theoretical): 84 kDa
Name EC 3.2.1, EC 3.2.1.31, KL, klotho
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Klotho protein Human Klotho (Glu34-His549) (N-terminus) Accession # Q9UEF7 Human Fractalkine Mucin-like Stalk (Phe103-Thr338) (N-terminus) Accession # P78423 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Klotho
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.