Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Kynurenine 3-Monooxygenase/KMO Protein
Shop All Novus Biologicals Products

Description
Measured by the consumption of NADPH by hydroxylation of L-kynurenine to 3-hydroxy-kynurenine. The specific activity is >290 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | O15229 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Brij-35. |
| Gene ID (Entrez) | 8564 |
| Molecular Weight (g/mol) | M.W. (Observed): 40-45 kDa, reducing conditions; M.W. (theoretical): 51 kDa |
| Name | Kynurenine 3-Monooxygenase/KMO |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Kynurenine 3-Monooxygenase/KMO protein Asp2-Leu441, with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?