Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LAG-3 Fc Chimera Protein
SDP

Catalog No. 2319L3050 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2319L3050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2319L3050 Supplier R&D Systems™ Supplier No. 2319L3050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce TNF-alpha secretion by JAWSII mouse immature dendritic cells. The ED50 for this effect is 1-4 μg/mL.

Specifications

Accession Number P18627
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3902
Molecular Weight (g/mol) 75-90 kDa, under reducing conditions.
Name LAG-3 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human LAG-3 protein Human LAG-3 (Leu23-Leu450) (N-terminus) Accession # P18627 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol LAG3
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS. Reconstitute 30 minutes prior to use with minimal agitation.
Product Line R&D Systems
Purity or Quality Grade Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, ≤ -20 °C as supplied. 1 month, 2 °C to 8 °C under sterile conditions after reconstitution. 3 months, ≤ -20 °C under sterile conditions after reconstitution.
Protein LAG-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.