Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Langerin/CD207 Fc Chimera Protein

Catalog No. 10401LN050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10401LN050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10401LN050 Supplier R&D Systems™ Supplier No. 10401LN050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human Langerin/CD207 Fc Chimera (Catalog # 10401-LN) is immobilized at 0.2 μg/mL (100 μL/well), Biotinylated Mannose-Polyacrylamide binds with an ED50 of 20-160 ng/mL.

Specifications

Accession Number Q9UJ71.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose.
Gene ID (Entrez) 50489
Molecular Weight (g/mol) M.W.-observed: 58-71 kDa, under reducing conditions, M.W.-theoretical: 56 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Langerin/CD207 protein MD Human IgG1 (Pro100-Lys330) IEGR Human Langerin/CD207 (Pro65-Pro328) (N-terminus) Accession # Q9UJ71.2 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD207, CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Langerin/CD207
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.