Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Latent BMP-9 Protein

Catalog No. 9624BP025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
96-24B-P025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 96-24B-P025 Supplier R&D Systems™ Supplier No. 9624BP025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Latent BMP-9 Protein is derived from CHO. The Recombinant Human Latent BMP-9 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9UK05
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 2658
Molecular Weight (g/mol) 33 kDa (pro) and 12 kDa (mature)
Name BMP9, BMP9BMP-9Bone morphogenetic protein 9, GDF2, GDF-2, growth differentiation factor 2, growth/differentiation factor 2
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human BMP-9 protein Lys23-Arg319 (pro) & Ser320-Arg429 (mature)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.