Learn More
R&D Systems™ Recombinant Human Latent TGF-beta 1 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein. |
| Gene ID (Entrez) | 7040 |
| Molecular Weight (g/mol) | Observed Molecular Wt: 40-43kDa & 12kDa, reducing conditions 80-90kDa, non-reducing conditions, Predicted Molecular Wt.: 41.3kDa (monomer) |
| Name | Latent TGF-beta 1 |
| Quantity | 5 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 1 protein Leu30-Ser390 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, ≤ -20° C under sterile conditions after opening. |
| Termination | Leu30 & Ala279 |
| Endotoxin Concentration | <1.0 EU per 1μg of the protein by the LAL method. |
| Gene Alias | CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.